Please login to your online account to display your discounted pricing

Mouse anti-CD27/TNFRSF7, Clone: O323, PE-Cy7, Novus Biologicals™

$197.00 - $362.00

Specifications

Antigen CD27/TNFRSF7
Clone O323
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD27.
Gene ID (Entrez) 939
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP142962

 
novus biologicals canada
NBP142962100TESTS
100 Tests Each for $362.00

NBP14296225

 
novus biologicals canada
NBP14296225TESTS
25 Tests Each for $197.00
Description & Specifications

Specifications

Antigen CD27/TNFRSF7
Clone O323
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD27.
Gene ID (Entrez) 939
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Murine
Isotype IgG1, κ
Conjugate PE-Cy7
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry