Please login to your online account to display your discounted pricing

Mouse anti-CD27/TNFRSF7, Clone: O323, PE-Cy5, Novus Biologicals™

$197.00 - $362.00

Specifications

Antigen CD27/TNFRSF7
Clone O323
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD27.
Gene ID (Entrez) 939
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP143092

 
novus biologicals canada
NBP143092100TESTS
100 Tests Each for $362.00

NBP14309225

 
novus biologicals canada
NBP14309225TESTS
25 Tests Each for $197.00
Description & Specifications

Specifications

Antigen CD27/TNFRSF7
Clone O323
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD27.
Gene ID (Entrez) 939
Formulation PBS [pH 7.2] containing BSA and/or gelatin for protein stabilization
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Murine
Isotype IgG1, κ
Conjugate PE-Cy5
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry