Please login to your online account to display your discounted pricing

Mouse anti-CD27/TNFRSF7, Clone: O323, FITC, Novus Biologicals™

$131.00 - $252.00

Specifications

Antigen CD27/TNFRSF7
Clone O323
Immunogen The immunogen for this antibody was CD27.
Gene ID (Entrez) 939
Formulation Phosphate buffer pH 7.2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP144020

 
novus biologicals canada
NBP144020100TESTS
100 Tests Each for $252.00

NBP14402025

 
novus biologicals canada
NBP14402025TESTS
25 Tests Each for $131.00
Description & Specifications

Specifications

Antigen CD27/TNFRSF7
Clone O323
Immunogen The immunogen for this antibody was CD27.
Gene ID (Entrez) 939
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate FITC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry