Please login to your online account to display your discounted pricing

Mouse anti-CD27/TNFRSF7, Clone: O323, Biotin, Novus Biologicals™

$131.00 - $252.00

Specifications

Antigen CD27/TNFRSF7
Clone O323
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD27.
Gene ID (Entrez) 939
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14366001

 
novus biologicals canada
NBP14366001MG
0.1mg Each for $252.00

NBP14366003

 
novus biologicals canada
NBP1436600025MG
0.025mg Each for $131.00
Description & Specifications

Specifications

Antigen CD27/TNFRSF7
Clone O323
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD27.
Gene ID (Entrez) 939
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Murine
Isotype IgG1, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry