Please login to your online account to display your discounted pricing

Mouse anti-CD25/IL-2 R alpha, Clone: BC96, Novus Biologicals™

$219.00 - $285.00

Specifications

Antigen CD25/IL-2 R alpha
Clone BC96
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD25.
Gene ID (Entrez) 3559
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14343001

 
novus biologicals canada
NBP14343001MG
0.1mg Each for $285.00

NBP14343003

 
novus biologicals canada
NBP1434300025MG
0.025mg Each for $219.00
Description & Specifications

Specifications

Antigen CD25/IL-2 R alpha
Clone BC96
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD25.
Gene ID (Entrez) 3559
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry