Please login to your online account to display your discounted pricing

Mouse anti-CD25/IL-2 R alpha, Clone: BC96, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1434300025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14343003

  • / Each
 

Description & Specifications

Specifications

Antigen CD25/IL-2 R alpha
Format Affinity Purified
Host Species Mouse
Immunogen The immunogen for this antibody was CD25.
Isotype IgG1, κ
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation Phosphate buffer pH 7.2
Applications Flow Cytometry
Purification Method Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.
Gene ID (Entrez) 3559
Clone BC96
Cross Reactivity Human
Concentration 0.5mg/mL
Conjugate Unlabeled