Please login to your online account to display your discounted pricing

Mouse anti-CD25/IL-2 R alpha, Clone: BC96, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14343001MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14343001

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Immunogen The immunogen for this antibody was CD25.
Host Species Mouse
Conjugate Unlabeled
Cross Reactivity Human
Quantity 0.1mg
Clone BC96
Primary or Secondary Primary
Gene ID (Entrez) 3559
Isotype IgG1, κ
Antigen CD25/IL-2 R alpha
Format Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.
Formulation Phosphate buffer pH 7.2
Regulatory Status RUO
Purification Method Affinity Purified
Monoclonal or Polyclonal Monoclonal
Concentration 0.5mg/mL