Please login to your online account to display your discounted pricing

Mouse anti-CD25/IL-2 R alpha, Clone: BC96, APC, Novus Biologicals™

$197.00 - $362.00

Specifications

Antigen CD25/IL-2 R alpha
Clone BC96
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD25.
Gene ID (Entrez) 3559
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP143049

 
novus biologicals canada
NBP143049100TESTS
100 Tests Each for $362.00

NBP14304925

 
novus biologicals canada
NBP14304925TESTS
25 Tests Each for $197.00
Description & Specifications

Specifications

Antigen CD25/IL-2 R alpha
Clone BC96
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD25.
Gene ID (Entrez) 3559
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate APC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry