Please login to your online account to display your discounted pricing

Mouse anti-CD24, Clone: SN3, Biotin, Novus Biologicals™

$96.00 - $192.00

Specifications

Antigen CD24
Clone SN3
Concentration 0.5mg/mL
Immunogen Glycoproteins purified from human NALM-1 cell line
Gene Accession No. P25063
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14366401

 
novus biologicals canada
NBP14366401MG
0.1mg Each for $192.00

NBP14366403

 
novus biologicals canada
NBP1436640025MG
0.025mg Each for $96.00
Description & Specifications

Specifications

Antigen CD24
Clone SN3
Concentration 0.5mg/mL
Immunogen Glycoproteins purified from human NALM-1 cell line
Gene Accession No. P25063
Gene ID (Entrez) 100133941
Formulation PBS [pH 7.2]. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Biotin
Format Purified
Purification Method Purified
Applications Flow Cytometry