Please login to your online account to display your discounted pricing

Mouse anti-CD24, Clone: SN3 A5-2H10, Novus Biologicals™

$219.00 - $285.00

Specifications

Antigen CD24
Clone SN3 A5-2H10
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD24.
Gene ID (Entrez) 100133941
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14343201

 
novus biologicals canada
NBP14343201MG
0.1mg Each for $285.00

NBP14343203

 
novus biologicals canada
NBP1434320025MG
0.025mg Each for $219.00
Description & Specifications

Specifications

Antigen CD24
Clone SN3 A5-2H10
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD24.
Gene ID (Entrez) 100133941
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry