Please login to your online account to display your discounted pricing

Mouse anti-CD24, Clone: SN3 A5-2H10, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1434320025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14343203

  • / Each
 

Description & Specifications

Specifications

Antigen CD24
Applications Flow Cytometry
Clone SN3 A5-2H10
Concentration 0.5mg/mL
Conjugate Unlabeled
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Host Species Murine
Immunogen The immunogen for this antibody was CD24.
Isotype IgG1
Purification Method Affinity Purified
Quantity 0.025mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 100133941
Target Species Human