Please login to your online account to display your discounted pricing

Mouse anti-CD24, Clone: SN3 A5-2H10, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14343201MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14343201

  • / Each
 

Description & Specifications

Specifications

Antigen CD24
Cross Reactivity Human
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Host Species Mouse
Applications Flow Cytometry
Immunogen The immunogen for this antibody was CD24.
Isotype IgG1
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Gene ID (Entrez) 100133941
Clone SN3 A5-2H10
Concentration 0.5mg/mL
Conjugate Unlabeled
Purification Method Affinity Purified
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal