Please login to your online account to display your discounted pricing

Mouse anti-CD2, Clone: UMCD2, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen CD2
Clone UMCD2
Concentration 1mg/mL
Immunogen Human Thymocytes/Sezary T cells
Gene Accession No. P06729
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23450801

 
novus biologicals canada
NBP23450801MG
0.1mg Each for $384.00

NBP23450802

 
novus biologicals canada
NBP23450802MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen CD2
Clone UMCD2
Concentration 1mg/mL
Immunogen Human Thymocytes/Sezary T cells
Gene Accession No. P06729
Gene ID (Entrez) 914
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG2a, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blot