Please login to your online account to display your discounted pricing

Mouse anti-CD2, Clone: UMCD2, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23450802MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23450802

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. P06729
Applications ELISA
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Conjugate Unlabeled
Host Species Mouse
Isotype IgG2a, κ
Purification Method Purified
Gene ID (Entrez) 914
Clone UMCD2
Antigen CD2
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Immunogen Human Thymocytes/Sezary T cells
Format Purified
Quantity 0.2mg
Concentration 1mg/mL
Primary or Secondary Primary