Please login to your online account to display your discounted pricing

Mouse anti-CD2, Clone: UMCD2, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23450801MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23450801

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Human Thymocytes/Sezary T cells
Isotype IgG2a, κ
Quantity 0.1mg
Primary or Secondary Primary
Gene ID (Entrez) 914
Cross Reactivity Human
Antigen CD2
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P06729
Host Species Mouse
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Clone UMCD2
Concentration 1mg/mL
Conjugate Unlabeled