Please login to your online account to display your discounted pricing

Mouse anti-CD2, Clone: BH1, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen CD2
Clone BH1
Concentration 1mg/mL
Immunogen Human CD2 protein
Gene Accession No. P06729
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23456102

 
novus biologicals canada
NBP23456102MG
0.2mg Each for $527.00

NBP23456101

 
novus biologicals canada
NBP23456101MG
0.1mg Each for $384.00
Description & Specifications

Specifications

Antigen CD2
Clone BH1
Concentration 1mg/mL
Immunogen Human CD2 protein
Gene Accession No. P06729
Gene ID (Entrez) 914
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2b, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation