Please login to your online account to display your discounted pricing

Mouse anti-CD2, Clone: BH1, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23456101MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23456101

  • / Each
 

Description & Specifications

Specifications

Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene ID (Entrez) 914
Cross Reactivity Human
Antigen CD2
Format Purified
Gene Accession No. P06729
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Immunogen Human CD2 protein
Isotype IgG2b, κ
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Concentration 1mg/mL
Purification Method Purified
Quantity 0.1mg
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Clone BH1
Conjugate Unlabeled