missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-CD2, Clone: 1E7E8.G4, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen CD2
Clone 1E7E8.G4
Concentration 1mg/mL
Immunogen Human Thymocytes/Sezary T cells
Gene Accession No. P06729
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23452301 Novus Biologicals Canada
NBP23452301MG
0.1mg Each for $384.00
NBP23452302 Novus Biologicals Canada
NBP23452302MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

CD2
1mg/mL
P06729
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
1E7E8.G4
Human Thymocytes/Sezary T cells
914
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG2a, κ
Purified
ELISA,Flow Cytometry,Functional Assay,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunoprecipitation,Western Blot