missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-CD1d, Clone: 51.1, Novus Biologicals™

$219.00 - $285.00

Specifications

Antigen CD1d
Clone 51.1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD1d.
Gene ID (Entrez) 912
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14346001 Novus Biologicals Canada
NBP14346001MG
0.1mg Each for $285.00
NBP14346003 Novus Biologicals Canada
NBP1434600025MG
0.025mg Each for $219.00
Specifications Description & Specifications

Specifications

CD1d
0.5mg/mL
912
Monoclonal
Store at 4°C. Do not freeze.
IgG2b
Affinity Purified
Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Frozen),Immunoprecipitation
51.1
The immunogen for this antibody was CD1d.
PBS [pH 7.2]
Primary
Murine
Unlabeled
Affinity Purified