Please login to your online account to display your discounted pricing

Mouse anti-CD1c/BDCA-1, Clone: L161, FITC, Novus Biologicals™

$142.00 - $296.00

Specifications

Antigen CD1c/BDCA-1
Clone L161
Immunogen Thymus cells
Gene Accession No. P29017
Gene ID (Entrez) 911
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP200068

 
novus biologicals canada
NBP200068100TESTS
100 Tests Each for $296.00

NBP20006825

 
novus biologicals canada
NBP20006825TESTS
25 Tests Each for $142.00
Description & Specifications

Specifications

Antigen CD1c/BDCA-1
Clone L161
Immunogen Thymus cells
Gene Accession No. P29017
Gene ID (Entrez) 911
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate FITC
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation