Please login to your online account to display your discounted pricing

Mouse anti-CD1b, Clone: SN13, Novus Biologicals™

$230.00 - $285.00

Specifications

Antigen CD1b
Clone SN13
Immunogen The immunogen for this antibody was CD1b.
Gene ID (Entrez) 910
Formulation PBS [pH 7.2]
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14345901

 
novus biologicals canada
NBP14345901MG
0.1mg Each for $285.00

NBP14345903

 
novus biologicals canada
NBP1434590025MG
0.025mg Each for $230.00
Description & Specifications

Specifications

Antigen CD1b
Clone SN13
Immunogen The immunogen for this antibody was CD1b.
Gene ID (Entrez) 910
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation