Please login to your online account to display your discounted pricing

Mouse anti-CD1b, Clone: SN13, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14345901MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14345901

  • / Each
 

Description & Specifications

Specifications

Antigen CD1b
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Clone SN13
Conjugate Unlabeled
Format Affinity Purified
Formulation PBS [pH 7.2]
Host Species Murine
Immunogen The immunogen for this antibody was CD1b.
Isotype IgG1
Purification Method Affinity Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 910
Target Species Human