Please login to your online account to display your discounted pricing

Mouse anti-CD1b, Clone: RIV12, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen CD1b
Clone RIV12
Concentration 1mg/mL
Immunogen Human peripheral lymphocytes
Gene Accession No. P29016
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23471401

 
novus biologicals canada
NBP23471401MG
0.1mg Each for $384.00

NBP23471402

 
novus biologicals canada
NBP23471402MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen CD1b
Clone RIV12
Concentration 1mg/mL
Immunogen Human peripheral lymphocytes
Gene Accession No. P29016
Gene ID (Entrez) 910
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation