Please login to your online account to display your discounted pricing

Mouse anti-CD1b, Clone: RIV12, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23471401MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23471401

  • / Each
 

Description & Specifications

Specifications

Quantity 0.1mg
Immunogen Human peripheral lymphocytes
Antigen CD1b
Isotype IgG1, κ
Conjugate Unlabeled
Cross Reactivity Human
Monoclonal or Polyclonal Monoclonal
Applications ELISA
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Primary or Secondary Primary
Purification Method Purified
Concentration 1mg/mL
Host Species Mouse
Gene Accession No. P29016
Regulatory Status RUO
Format Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Reconstitution Protein A purified
Gene ID (Entrez) 910
Clone RIV12