Please login to your online account to display your discounted pricing

Mouse anti-CD1a, Clone: SPM120, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23301710ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23301710

  • / Each
 

Description & Specifications

Specifications

Formulation PBS with 0.05% BSA.
Gene Accession No. P06126
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 1.0mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Antigen CD1a
Format Purified
Isotype IgG1, κ
Purification Method Purified
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 909
Clone SPM120
Conjugate Unlabeled
Cross Reactivity Human
Immunogen Human thymus cells
Reconstitution Protein A purified
Concentration 0.2mg/mL