Please login to your online account to display your discounted pricing

Mouse anti-CD1a, Clone: SPM120, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23473101MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23473101

  • / Each
 

Description & Specifications

Specifications

Antigen CD1a
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Human thymus cells
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 909
Reconstitution Protein A purified
Clone SPM120
Concentration 1mg/mL
Cross Reactivity Human
Gene Accession No. P06126
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled