Please login to your online account to display your discounted pricing

Mouse anti-CD1a, Clone: SPM120, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23473102MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23473102

  • / Each
 

Description & Specifications

Specifications

Immunogen Human thymus cells
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P06126
Host Species Mouse
Isotype IgG1, κ
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 909
Reconstitution Protein A purified
Conjugate Unlabeled
Antigen CD1a
Clone SPM120
Concentration 1mg/mL