Please login to your online account to display your discounted pricing

Mouse anti-CD1a, Clone: O10+C1A/711, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23431405ML

 View all options for this product


For Research Use Only

Catalog No. NBP23431405


Description & Specifications


Antigen CD1a
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone O10+C1A/711
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P06126
Host Species Mouse
Immunogen Human thymus cells (O10); Recombinant human CD1a protein (C1A/711)
Isotype IgG1, κ
Purification Method Purified
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 909
Reconstitution Protein A purified