Please login to your online account to display your discounted pricing

Mouse anti-CD1a, Clone: O10+C1A/711, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen CD1a
Clone O10+C1A/711
Concentration 1mg/mL
Immunogen Human thymus cells (O10); Recombinant human CD1a protein (C1A/711)
Gene Accession No. P06126
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23469802

 
novus biologicals canada
NBP23469802MG
0.2mg Each for $527.00

NBP23469801

 
novus biologicals canada
NBP23469801MG
0.1mg Each for $384.00
Description & Specifications

Specifications

Antigen CD1a
Clone O10+C1A/711
Concentration 1mg/mL
Immunogen Human thymus cells (O10); Recombinant human CD1a protein (C1A/711)
Gene Accession No. P06126
Gene ID (Entrez) 909
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting