Please login to your online account to display your discounted pricing

Mouse anti-CD1a, Clone: O10+C1A/711, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23469801MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23469801

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen CD1a
Format Purified
Gene Accession No. P06126
Host Species Mouse
Isotype IgG1, κ
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Gene ID (Entrez) 909
Conjugate Unlabeled
Cross Reactivity Human
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Immunogen Human thymus cells (O10); Recombinant human CD1a protein (C1A/711)
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Clone O10+C1A/711
Concentration 1mg/mL