missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-CD1a, Clone: O10, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen CD1a
Clone O10
Concentration 1mg/mL
Immunogen Human thymus cells
Gene Accession No. P06126
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23469701 Novus Biologicals Canada
NBP23469701MG
0.1mg Each for $384.00
NBP23469702 Novus Biologicals Canada
NBP23469702MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

CD1a
1mg/mL
P06126
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
O10
Human thymus cells
909
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
ELISA,Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot