Please login to your online account to display your discounted pricing

Mouse anti-CD1a, Clone: O10, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23469702MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23469702

  • / Each
 

Description & Specifications

Specifications

Format Purified
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P06126
Immunogen Human thymus cells
Isotype IgG1, κ
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 909
Reconstitution Protein A purified
Clone O10
Concentration 1mg/mL
Conjugate Unlabeled
Antigen CD1a