Please login to your online account to display your discounted pricing

Mouse anti-CD1a, Clone: O10, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23469701MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23469701

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Format Purified
Host Species Mouse
Antigen CD1a
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Conjugate Unlabeled
Cross Reactivity Human
Reconstitution Protein A purified
Purification Method Purified
Clone O10
Gene Accession No. P06126
Gene ID (Entrez) 909
Monoclonal or Polyclonal Monoclonal
Quantity 0.1mg
Immunogen Human thymus cells
Primary or Secondary Primary
Isotype IgG1, κ
Concentration 1mg/mL
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Regulatory Status RUO