Please login to your online account to display your discounted pricing

Mouse anti-CD1a, Clone: HI149, Biotin, Novus Biologicals™

$131.00 - $252.00

Specifications

Antigen CD1a
Clone HI149
Concentration 0.5mg/mL
Immunogen Tissue / cell preparation (Human thymocytes).
Gene ID (Entrez) 909
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14369601

 
novus biologicals canada
NBP14369601MG
0.1mg Each for $252.00

NBP14369603

 
novus biologicals canada
NBP1436960025MG
0.025mg Each for $131.00
Description & Specifications

Specifications

Antigen CD1a
Clone HI149
Concentration 0.5mg/mL
Immunogen Tissue / cell preparation (Human thymocytes).
Gene ID (Entrez) 909
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry