Please login to your online account to display your discounted pricing

Mouse anti-CD1a, Clone: C1A/711, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23457401MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23457401

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Quantity 0.1mg
Gene ID (Entrez) 909
Regulatory Status RUO
Gene Accession No. P06126
Primary or Secondary Primary
Purification Method Purified
Isotype IgG1, κ
Immunogen Recombinant human CD1a protein
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Antigen CD1a
Reconstitution Protein A purified
Clone C1A/711
Concentration 1mg/mL
Host Species Mouse