Please login to your online account to display your discounted pricing

Mouse anti-CD19, Clone: SJ25-C1, Cy5.5, PerCP, Novus Biologicals™

$197.00 - $395.00

Specifications

Antigen CD19
Clone SJ25-C1
Immunogen The immunogens are the NALM-1 and NALM-16 human cell lines.
Gene ID (Entrez) 930
Formulation IgG purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP200018

 
novus biologicals canada
NBP200018100TESTS
100 Tests Each for $395.00

NBP20001825

 
novus biologicals canada
NBP20001825TESTS
25 Tests Each for $197.00
Description & Specifications

Specifications

Antigen CD19
Clone SJ25-C1
Immunogen The immunogens are the NALM-1 and NALM-16 human cell lines.
Gene ID (Entrez) 930
Formulation IgG purified
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Murine
Isotype IgG1
Conjugate Cy5.5, PerCP
Format Purified
Purification Method Purified
Applications Flow Cytometry