Please login to your online account to display your discounted pricing

Mouse anti-CD19, Clone: MB19-1, FITC, Novus Biologicals™

$192.00 - $192.00

Specifications

Antigen CD19
Clone MB19-1
Concentration 0.5mg/mL
Immunogen mouse CD19+ pre-B cell line 300.19
Gene ID (Entrez) 930
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP20002410

 
novus biologicals canada
NBP200024100UG
100μg Each for $192.00
Description & Specifications

Specifications

Antigen CD19
Clone MB19-1
Concentration 0.5mg/mL
Immunogen mouse CD19+ pre-B cell line 300.19
Gene ID (Entrez) 930
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Mouse
Isotype IgA, κ
Conjugate FITC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry