Please login to your online account to display your discounted pricing

Mouse anti-CD19, Clone: MB19-1, Biotin, Novus Biologicals™

$171.00 - $418.00

Specifications

Antigen CD19
Clone MB19-1
Concentration 0.2mg/mL
Immunogen mouse CD19+ pre-B cell line 300.19
Gene ID (Entrez) 930
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14367005

 
novus biologicals canada
NBP143670005MG
0.05mg Each for $171.00

NBP14367001

 
novus biologicals canada
NBP14367001MG
0.1mg Each for $192.00

NBP1436705

 
novus biologicals canada
NBP14367005MG
0.5mg Each for $418.00
Description & Specifications

Specifications

Antigen CD19
Clone MB19-1
Concentration 0.2mg/mL
Immunogen mouse CD19+ pre-B cell line 300.19
Gene ID (Entrez) 930
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Mouse
Isotype IgA, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry