Please login to your online account to display your discounted pricing

Mouse anti-CD19, Clone: HIB19, PE-Cy5, Novus Biologicals™

$175.00 - $329.00

Specifications

Antigen CD19
Clone HIB19
Immunogen The immunogen for this antibody was CD19.
Gene ID (Entrez) 930
Formulation Phosphate buffer pH 7.2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP143096

 
novus biologicals canada
NBP143096100TESTS
100 Tests Each for $329.00

NBP14309625

 
novus biologicals canada
NBP14309625TESTS
25 Tests Each for $175.00
Description & Specifications

Specifications

Antigen CD19
Clone HIB19
Immunogen The immunogen for this antibody was CD19.
Gene ID (Entrez) 930
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate PE-Cy5
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry