Please login to your online account to display your discounted pricing

Mouse anti-CD163, Clone: GHI/61, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1433410025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14334103

  • / Each
 

Description & Specifications

Specifications

Antigen CD163
Applications Flow Cytometry
Applications Western Blot
Clone GHI/61
Conjugate Unlabeled
Format Affinity Purified
Formulation PBS [pH 7.2]
Host Species Murine
Immunogen The immunogen for this antibody was CD163.
Isotype IgG1, κ
Purification Method Affinity Purified
Quantity 0.025mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 9332
Target Species Human