Please login to your online account to display your discounted pricing

Mouse anti-CD163, Clone: GHI/61, Biotin, Novus Biologicals™

$131.00 - $252.00

Specifications

Antigen CD163
Clone GHI/61
Immunogen The immunogen for this antibody was CD163.
Gene ID (Entrez) 9332
Formulation PBS [pH 7.2]
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14357603

 
novus biologicals canada
NBP1435760025MG
0.025mg Each for $131.00

NBP14357601

 
novus biologicals canada
NBP14357601MG
0.1mg Each for $252.00
Description & Specifications

Specifications

Antigen CD163
Clone GHI/61
Immunogen The immunogen for this antibody was CD163.
Gene ID (Entrez) 9332
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Murine
Isotype IgG1, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry