Please login to your online account to display your discounted pricing

Mouse anti-CD163, Clone: 6E10.1G6, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP236494SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP236494SS

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Applications Flow Cytometry
Applications Western Blotting
Cross Reactivity Human
Immunogen A synthetic peptide made to the human CD163 protein sequence (between residues 200-400). [Swiss-Prot Q86VB7]
Formulation Protein G purified
Regulatory Status RUO
Isotype IgG2b, κ
Clone 6E10.1G6
Format Purified
Quantity 0.025mg
Gene ID (Entrez) 9332
Antigen CD163
Concentration 1.0mg/mL
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Purification Method Purified
Test Specificity This antibody is specific for the extracellular domain of human CD163 protein. Immunogen's sequence similarity with other species: Bovine (88%), Porcine (87%), Canine (87%), Rat (80%), Mouse (78%)