Please login to your online account to display your discounted pricing

Mouse anti-CD163, Clone: 6D3.2F3, Novus Biologicals™

$109.00 - $307.00

Specifications

Antigen CD163
Clone 6D3.2F3
Concentration 1.0mg/mL
Immunogen A synthetic peptide made to the human CD163 protein sequence (between residues 200-400). [Swiss-Prot Q86VB7]
Gene Accession No. Q86VB7
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP236495

 
novus biologicals canada
NBP236495
0.1mg Each for $307.00

NBP236495SS

 
novus biologicals canada
NBP236495SS
0.025mg Each for $109.00
Description & Specifications

Specifications

Antigen CD163
Clone 6D3.2F3
Concentration 1.0mg/mL
Immunogen A synthetic peptide made to the human CD163 protein sequence (between residues 200-400). [Swiss-Prot Q86VB7]
Gene Accession No. Q86VB7
Gene ID (Entrez) 9332
Formulation PBS. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2b, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Western Blotting