Please login to your online account to display your discounted pricing

Mouse anti-CD161/NK1.1, Clone: PK136, Biotin, Novus Biologicals™

$128.00 - $386.00

Specifications

Antigen CD161/NK1.1
Clone PK136
Concentration 0.2mg/mL
Immunogen NK-1+ cells from mouse spleen and bone marrow
Gene ID (Entrez) 17059
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14351805

 
novus biologicals canada
NBP143518005MG
0.05mg Each for $128.00

NBP14351801

 
novus biologicals canada
NBP14351801MG
0.1mg Each for $192.00

NBP1435185

 
novus biologicals canada
NBP14351805MG
0.5mg Each for $386.00
Description & Specifications

Specifications

Antigen CD161/NK1.1
Clone PK136
Concentration 0.2mg/mL
Immunogen NK-1+ cells from mouse spleen and bone marrow
Gene ID (Entrez) 17059
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry