Please login to your online account to display your discounted pricing

Mouse anti-CD161/NK1.1, Clone: PK136, Biotin, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143518005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14351805

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Applications Flow Cytometry
Primary or Secondary Primary
Antigen CD161/NK1.1
Formulation Phosphate buffer pH 7.2
Immunogen NK-1+ cells from mouse spleen and bone marrow
Isotype IgG2a, κ
Purification Method Affinity Purified
Quantity 0.05mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 17059
Test Specificity Mouse Monoclonal to NK-1.1 (PK136)
Clone PK136
Concentration 0.2mg/mL
Conjugate Biotin
Cross Reactivity Mouse
Format Affinity Purified