Please login to your online account to display your discounted pricing

Mouse anti-CD161/NK1.1, Clone: PK136, Biotin, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14351805MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1435185

  • / Each
 

Description & Specifications

Specifications

Antigen CD161/NK1.1
Applications Flow Cytometry
Storage Requirements Store at 4°C. Do not freeze.
Format Affinity Purified
Host Species Mouse
Immunogen NK-1+ cells from mouse spleen and bone marrow
Isotype IgG2a, κ
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity Mouse Monoclonal to NK-1.1 (PK136)
Formulation Phosphate buffer pH 7.2
Purification Method Affinity Purified
Quantity 0.5mg
Clone PK136
Concentration 0.2mg/mL
Conjugate Biotin
Cross Reactivity Mouse
Gene ID (Entrez) 17059