Please login to your online account to display your discounted pricing

Mouse anti-CD161/NK1.1, Clone: PK136, Biotin, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14351801MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14351801

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Cross Reactivity Mouse
Regulatory Status RUO
Gene ID (Entrez) 17059
Immunogen NK-1+ cells from mouse spleen and bone marrow
Monoclonal or Polyclonal Monoclonal
Host Species Mouse
Format Affinity Purified
Purification Method Affinity Purified
Clone PK136
Concentration 0.2mg/mL
Isotype IgG2a, κ
Storage Requirements Store at 4°C. Do not freeze.
Test Specificity Mouse Monoclonal to NK-1.1 (PK136)
Antigen CD161/NK1.1
Quantity 0.1mg
Conjugate Biotin
Primary or Secondary Primary
Formulation Phosphate buffer pH 7.2