Please login to your online account to display your discounted pricing

Mouse anti-CD15/Lewis X, Clone: SPM119, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen CD15/Lewis X
Clone SPM119
Immunogen U937 histiocytic cell line
Gene Accession No. P22083
Gene ID (Entrez) 2526
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23444801

 
novus biologicals canada
NBP23444801ML
0.1mL Each for $139.00

NBP23444805

 
novus biologicals canada
NBP23444805ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen CD15/Lewis X
Clone SPM119
Immunogen U937 histiocytic cell line
Gene Accession No. P22083
Gene ID (Entrez) 2526
Formulation Culture Medium. Bioreactor concentrate.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgM, κ
Conjugate Unlabeled
Format Concentrated
Purification Method Concentrated
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence