Please login to your online account to display your discounted pricing

Mouse anti-CD15/Lewis X, Clone: SPM119, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23444805ML

 View all options for this product


For Research Use Only

Catalog No. NBP23444805


Description & Specifications


Antigen CD15/Lewis X
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Clone SPM119
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Formulation Culture Medium. Bioreactor concentrate.
Gene Accession No. P22083
Host Species Mouse
Immunogen U937 histiocytic cell line
Isotype IgM, κ
Purification Method Concentrated
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2526