Please login to your online account to display your discounted pricing

Mouse anti-CD15/Lewis X, Clone: SPM119, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23444801ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23444801

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Antigen CD15/Lewis X
Format Concentrated
Formulation Culture Medium. Bioreactor concentrate.
Gene Accession No. P22083
Quantity 0.1mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 2526
Conjugate Unlabeled
Cross Reactivity Human
Host Species Mouse
Immunogen U937 histiocytic cell line
Isotype IgM, κ
Purification Method Concentrated
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone SPM119